Please login to your online account to display your discounted pricing

Rat IgG1 Kappa Light Chain, Clone: RG1, Biotin, Isotype control, Novus Biologicals™

$219.00 - $736.00

Specifications

Antigen IgG1 Kappa Light Chain
Primary or Secondary Primary
Host Species Rat
Conjugate Biotin
Applications ELISA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14356201

 
novus biologicals canada
NBP14356201MG
0.1mg Each for $329.00

NBP14356205

 
novus biologicals canada
NBP143562005MG
0.05mg Each for $219.00

NBP1435625

 
novus biologicals canada
NBP14356205MG
0.5mg Each for $736.00
Description & Specifications

Specifications

Antigen IgG1 Kappa Light Chain
Primary or Secondary Primary
Host Species Rat
Conjugate Biotin
Applications ELISA
Applications Flow Cytometry
Storage Requirements Store at 4°C. Do not freeze.