Please login to your online account to display your discounted pricing

Rat IgG1 Kappa Light Chain, Clone: RG1, APC, Isotype control, Novus Biologicals™

$252.00 - $549.00

Specifications

Antigen IgG1 Kappa Light Chain
Primary or Secondary Primary
Host Species Rat
Conjugate Allophycocyanin
Applications Flow Cytometry
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14300601

 
novus biologicals canada
NBP14300601MG
0.1mg Each for $362.00

NBP14300602

 
novus biologicals canada
NBP14300602MG
0.2mg Each for $549.00

NBP14300605

 
novus biologicals canada
NBP143006005MG
0.05mg Each for $252.00
Description & Specifications

Specifications

Antigen IgG1 Kappa Light Chain
Primary or Secondary Primary
Host Species Rat
Conjugate Allophycocyanin
Applications Flow Cytometry
Storage Requirements Store at 4°C. Do not freeze.