missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Rat IgG1 ELISA Ready-SET-Go!™ Kit

For quantitative detection of rat IgG1.

Manufacturer:  Invitrogen™ LS885050077

 View more versions of this product

Catalog No. 501125133



Includes: Capture Antibody Concentrate
Detection Antibody Concentrate
Rat IgG1 isotype control (standard), lyophilized, 800ng/mL upon reconstitution
Phosphate Buffered Saline Concentrate (PBS)
Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)

Description

Description

Rat IgG1 Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications

Specifications

ELISA
6.25ng/mL
20μL (prediluted 1:10,000)
2° to 8°C
Immunoglobulin G
20 x 96 tests
Cell culture supernatant, serum and plasma
Rat
IgG1
Documents

For Research Use Only