Please login to your online account to display your discounted pricing

eBioscience™ Rat IgA ELISA Ready-SET-Go!™ Kit

For quantitative detection of rat IgA.

$332.05 - $999.05

Specifications

For Use With (Application) ELISA
Sample Size 50μL (prediluted 1:1,000)
Synonym Immunoglobulin A
Target IgA
Sample Type Cell culture supernatant, serum and plasma
View More Specs
Includes:  Capture Antibody Concentrate

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

501125122

 
life technologies
LS885048022
96-well plates 2 x 96 tests Each for $332.05

501125123

 
life technologies
LS885048076
96-well plates 20 x 96 tests Each for $999.05

501125125

 
life technologies
LS885048086
96-well plates 10 x 96 tests Each for $694.55

501125124

 
life technologies
LS885048077
Each for $941.05

501125126

 
life technologies
LS885048088
Each for $665.55
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL (prediluted 1:1,000)
Synonym Immunoglobulin A
Target IgA
Sample Type Cell culture supernatant, serum and plasma
Sensitivity 16ng/mL
Storage Requirements 2° to 8°C
Species Rat

Rat IgA Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)