Invitrogen™ eBioscience™ Rat IgA ELISA Ready-SET-Go!™ Kit
For quantitative detection of rat IgA.
Manufacturer:  Invitrogen™ LS885048086
 View more versions of this product
Catalog No. 501125125
Includes: Capture Antibody Concentrate
Description
Description
Rat IgA Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications
Specifications
ELISA
10 x 96 tests
Cell culture supernatant, serum and plasma
Rat
IgA
96-well plates
16ng/mL
50μL (prediluted 1:1,000)
2° to 8°C
Immunoglobulin A