Please login to your online account to display your discounted pricing

eBioscience™ Rat Ig Isotyping ELISA Ready-SET-Go!™ Kit

Specifically engineered for measurement of rat Immunoglobulins from cell culture supernatants. Up to 12 individual samples can be tested for anti-IgG1, IgG2a, IgG2b, IgG2c, IgA, and IgM as well as light chain kappa and lambda.

$723.55 - $752.55

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Quantity 10 x 96 tests
Target Ig kappa light chain, Ig lambda light chain, IgA, IgG1, IgG2a, IgG2b, IgG2c, IgM
Sample Type Cell culture supernatant
View More Specs
Includes:  Capture Antibody Concentrate
Detection Antibody Concentrate
Rat Ig isotype control mixture (positive control), lyophilized
Phosphate Buffered Saline Concentrate (PBS)
Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Includes Price Quantity    

501125160

 
life technologies
LS885064088
Each for $723.55

501125159

 
life technologies
LS885064086
96-well plates Each for $752.55
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Quantity 10 x 96 tests
Target Ig kappa light chain, Ig lambda light chain, IgA, IgG1, IgG2a, IgG2b, IgG2c, IgM
Sample Type Cell culture supernatant
Species Rat

Rat Ig Isotyping Ready-SET-Go! ELISA Set contains the necessary reagents, controls, buffers and diluents for performing qualititative enzyme-linked immunosorbent assays (ELISA)