missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat IFN-gamma Quantikine ELISA Kit, R&D Systems

a 4.5 hour solid phase ELISA designed to measure rat IFN-gamma levels in cell culture supernates and serum.

$606.82 - $2,912.46

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 25712
Sample Type Cell Culture Supernates, Serum
Sample Volume Cell Culture Supernates (50μL), Serum (50μL)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
RIF00 R & D Systems
RDSRIF00
1 Plate Each for $606.82
SRIF00 R & D Systems
RDSSRIF00
6 Plates Each for $2,912.46
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL), Serum (50μL)
Sensitivity: 10pg/mL
Specificity: Natural and recombinant rat IFN-gamma
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
25712
Cell Culture Supernates (50μL), Serum (50μL)
Rat
Solid Phase Sandwich ELISA
Cell Culture Supernates, Serum
10pg/mL
Natural and recombinant rat IFN-gamma