Please login to your online account to display your discounted pricing

Rat anti-VEGF R3/Flt-4, Clone: AFL4, Novus Biologicals™

$219.00 - $362.00

Specifications

Antigen VEGF R3/Flt-4
Clone AFL4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was VEGFR3.
Gene ID (Entrez) 2324
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14325901

 
novus biologicals canada
NBP14325901MG
0.1mg Each for $285.00

NBP14325905

 
novus biologicals canada
NBP143259005MG
0.05mg Each for $219.00

NBP1432595

 
novus biologicals canada
NBP14325905MG
0.5mg Each for $362.00
Description & Specifications

Specifications

Antigen VEGF R3/Flt-4
Clone AFL4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was VEGFR3.
Gene ID (Entrez) 2324
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blot