Please login to your online account to display your discounted pricing

Rat anti-VEGF R3/Flt-4, Clone: AFL4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143259005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14325905

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Rat
Conjugate Unlabeled
Regulatory Status RUO
Isotype IgG2a, κ
Cross Reactivity Mouse
Immunogen The immunogen for this antibody was VEGFR3.
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Antigen VEGF R3/Flt-4
Monoclonal or Polyclonal Monoclonal
Quantity 0.05mg
Clone AFL4
Gene ID (Entrez) 2324
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Concentration 0.5mg/mL
Purification Method Affinity Purified