Please login to your online account to display your discounted pricing

Rat anti-VEGF R3/Flt-4, Clone: AFL4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14325905MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1432595

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Mouse
Antigen VEGF R3/Flt-4
Immunogen The immunogen for this antibody was VEGFR3.
Purification Method Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone AFL4
Conjugate Unlabeled
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Rat
Isotype IgG2a, κ
Quantity 0.5mg
Gene ID (Entrez) 2324
Concentration 0.5mg/mL