Please login to your online account to display your discounted pricing

Rat anti-VEGF R3/Flt-4, Clone: AFL4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14325901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14325901

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Antigen VEGF R3/Flt-4
Conjugate Unlabeled
Formulation Phosphate buffer pH 7.2
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Concentration 0.5mg/mL
Gene ID (Entrez) 2324
Purification Method Affinity Purified
Isotype IgG2a, κ
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Mouse
Clone AFL4
Host Species Rat
Primary or Secondary Primary
Format Affinity Purified
Immunogen The immunogen for this antibody was VEGFR3.
Quantity 0.1mg