missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-VEGF R3/Flt-4, Clone: AFL4, Biotin, Novus Biologicals™

$252.00 - $329.00

Specifications

Antigen VEGF R3/Flt-4
Clone AFL4
Immunogen The immunogen for this antibody was VEGFR3.
Gene ID (Entrez) 2324
Formulation Phosphate buffer pH 7.2
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14350201 Novus Biologicals Canada
NBP14350201MG
0.1mg Each for $329.00
NBP14350205 Novus Biologicals Canada
NBP143502005MG
0.05mg Each for $252.00
Specifications Description & Specifications

Specifications

VEGF R3/Flt-4
The immunogen for this antibody was VEGFR3.
Phosphate buffer pH 7.2
Primary
Rat
Biotin
Affinity Purified
AFL4
2324
Monoclonal
Store at 4°C. Do not freeze.
IgG2a, κ
Affinity Purified
Flow Cytometry