missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-VEGF R2/KDR/Flk-1, Clone: Avas12a1, Novus Biologicals™

$230.00 - $362.00

Specifications

Antigen VEGF R2/KDR/Flk-1
Clone Avas12a1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Flk1.
Gene ID (Entrez) 3791
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14330001 Novus Biologicals Canada
NBP14330001MG
0.1mg Each for $285.00
NBP14330005 Novus Biologicals Canada
NBP143300005MG
0.05mg Each for $230.00
NBP1433005 Novus Biologicals Canada
NBP14330005MG
0.5mg Each for $362.00
Specifications Description & Specifications

Specifications

VEGF R2/KDR/Flk-1
0.5mg/mL
3791
Monoclonal
Store at 4°C. Do not freeze.
IgG2a, κ
Affinity Purified
Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot
Avas12a1
The immunogen for this antibody was Flk1.
Phosphate buffer pH 7.2
Primary
Rat
Unlabeled
Affinity Purified