Please login to your online account to display your discounted pricing

Rat anti-VEGF R2/KDR/Flk-1, Clone: Avas12a1, Biotin, Novus Biologicals™

$252.00 - $725.00

Specifications

Antigen VEGF R2/KDR/Flk-1
Clone Avas12a1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Flk1.
Gene ID (Entrez) 3791
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14354301

 
novus biologicals canada
NBP14354301MG
0.1mg Each for $329.00

NBP14354305

 
novus biologicals canada
NBP143543005MG
0.05mg Each for $252.00

NBP1435435

 
novus biologicals canada
NBP14354305MG
0.5mg Each for $725.00
Description & Specifications

Specifications

Antigen VEGF R2/KDR/Flk-1
Clone Avas12a1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Flk1.
Gene ID (Entrez) 3791
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry