Please login to your online account to display your discounted pricing

Rat anti-VE-Cadherin, Clone: BV14, Novus Biologicals™

$246.00 - $406.00

Specifications

Antigen VE-Cadherin
Clone BV14
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD144.
Gene ID (Entrez) 1003
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14334701

 
novus biologicals canada
NBP14334701MG
0.1mg Each for $307.00

NBP14334705

 
novus biologicals canada
NBP143347005MG
0.05mg Each for $246.00

NBP1433475

 
novus biologicals canada
NBP14334705MG
0.5mg Each for $406.00
Description & Specifications

Specifications

Antigen VE-Cadherin
Clone BV14
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD144.
Gene ID (Entrez) 1003
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting