Please login to your online account to display your discounted pricing

Rat anti-VE-Cadherin, Clone: BV14, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14334705MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1433475

  • / Each
 

Description & Specifications

Specifications

Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Formulation PBS [pH 7.2]
Host Species Rat
Purification Method Affinity Purified
Primary or Secondary Primary
Gene ID (Entrez) 1003
Conjugate Unlabeled
Antigen VE-Cadherin
Format Affinity Purified
Immunogen The immunogen for this antibody was CD144.
Isotype IgG2b
Quantity 0.5mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Clone BV14
Concentration 0.5mg/mL
Cross Reactivity Mouse