missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-VE-Cadherin, Clone: BV13, Novus Biologicals™

$252.00 - $384.00

Specifications

Antigen VE-Cadherin
Clone BV13
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD144.
Gene Accession No. P33151
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14334801 Novus Biologicals Canada
NBP14334801MG
0.1mg Each for $329.00
NBP14334805 Novus Biologicals Canada
NBP143348005MG
0.05mg Each for $252.00
NBP1433485 Novus Biologicals Canada
NBP14334805MG
0.5mg Each for $384.00
Specifications Description & Specifications

Specifications

VE-Cadherin
0.5mg/mL
P33151
PBS [pH 7.2]
Primary
Rat
Unlabeled
Affinity Purified
BV13
The immunogen for this antibody was CD144.
1003
Monoclonal
Store at 4°C. Do not freeze.
IgG1
Affinity Purified
Flow Cytometry,Functional Assay,Immunohistochemistry,Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot