Please login to your online account to display your discounted pricing

Rat anti-VE-Cadherin, Clone: BV13, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14334801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14334801

  • / Each
 

Description & Specifications

Specifications

Antigen VE-Cadherin
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blot
Clone BV13
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation PBS [pH 7.2]
Gene Accession No. P33151
Host Species Rat
Immunogen The immunogen for this antibody was CD144.
Isotype IgG1
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1003
Target Species Murine