Please login to your online account to display your discounted pricing

Rat anti-VE-Cadherin, Clone: BV13, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143348005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14334805

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen The immunogen for this antibody was CD144.
Antigen VE-Cadherin
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Rat
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1003
Clone BV13
Cross Reactivity Mouse
Gene Accession No. P33151
Isotype IgG1
Quantity 0.05mg
Regulatory Status RUO
Concentration 0.5mg/mL
Conjugate Unlabeled