Please login to your online account to display your discounted pricing

Rat anti-V alpha 2 TCR, Clone: B20.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200142500UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20014255

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunoprecipitation
Antigen V alpha 2 TCR
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is V alpha 2 TCR (Va2-TCR
Isotype IgG2a
Storage Requirements Store at 4°C. Do not freeze.
Regulatory Status RUO
Cross Reactivity Mouse
Conjugate Unlabeled
Host Species Rat
Purification Method Affinity Purified
Quantity 500μg
Format Affinity Purified
Clone B20.1