Please login to your online account to display your discounted pricing

Rat anti-V alpha 2 TCR, Clone: B20.1, Biotin, Novus Biologicals™

$329.00 - $725.00

Specifications

Antigen V alpha 2 TCR
Clone B20.1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is V alpha 2 TCR (Va2-TCR
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20014110

 
novus biologicals canada
NBP200141100UG
100μg Each for $329.00

NBP20014155

 
novus biologicals canada
NBP200141500UG
500μg Each for $725.00
Description & Specifications

Specifications

Antigen V alpha 2 TCR
Clone B20.1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is V alpha 2 TCR (Va2-TCR
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunoprecipitation