Please login to your online account to display your discounted pricing

Rat anti-TWEAK/TNFSF12, Clone: MTW-1, Biotin, Novus Biologicals™

$192.00 - $493.00

Specifications

Antigen TWEAK/TNFSF12
Clone MTW-1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was TWEAK.
Gene ID (Entrez) 8742
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14347601

 
novus biologicals canada
NBP14347601MG
0.1mg Each for $246.00

NBP1434765

 
novus biologicals canada
NBP14347605MG
0.5mg Each for $493.00

NBP14347605

 
novus biologicals canada
NBP143476005MG
0.05mg Each for $192.00
Description & Specifications

Specifications

Antigen TWEAK/TNFSF12
Clone MTW-1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was TWEAK.
Gene ID (Entrez) 8742
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG1, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry