missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-TREM-1, Clone: L5-B8, Novus Biologicals™

$309.33 - $479.90

Specifications

Antigen TREM-1
Clone L5-B8
Concentration 0.1mg/mL
Immunogen Mouse TREM-1-human IgG-Fc fusion protein
Gene ID (Entrez) 54210
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14960601 Novus Biologicals Canada
NBP14960601MG
0.1mg Each for $479.90
NBP14960605 Novus Biologicals Canada
NBP149606005MG
0.05mg Each for $309.33
Specifications Description & Specifications

Specifications

TREM-1
0.1mg/mL
54210
Monoclonal
Store at 4°C. Do not freeze.
IgG2a
Affinity Purified
Flow Cytometry,Functional Assay,Immunocytochemistry,Immunofluorescence,Western Blot
L5-B8
Mouse TREM-1-human IgG-Fc fusion protein
1mL (100μg/mL) 0.2μm filtered antibody solution in PBS, containing 0.1% bovine serum albumin.
Primary
Rat
Unlabeled
Affinity Purified