Please login to your online account to display your discounted pricing

Rat anti-TREM-1, Clone: L5-B8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP149606005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14960605

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Mouse
Format Affinity Purified
Formulation 1mL (100μg/mL) 0.2μm filtered antibody solution in PBS, containing 0.1% bovine serum albumin.
Host Species Rat
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Isotype IgG2a
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Mouse TREM-1
Antigen TREM-1
Immunogen Mouse TREM-1-human IgG-Fc fusion protein
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 54210
Clone L5-B8
Concentration 0.1mg/mL
Conjugate Unlabeled