Please login to your online account to display your discounted pricing

Rat anti-TREM-1, Clone: L5-B8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14960601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14960601

  • / Each
 

Description & Specifications

Specifications

Antigen TREM-1
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Clone L5-B8
Concentration 0.1mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation 1mL (100μg/mL) 0.2μm filtered antibody solution in PBS, containing 0.1% bovine serum albumin.
Host Species Rat
Immunogen Mouse TREM-1-human IgG-Fc fusion protein
Isotype IgG2a
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 54210
Test Specificity Mouse TREM-1
Target Species Murine