Please login to your online account to display your discounted pricing

Rat anti-TRANCE/TNFSF11/RANK L, Clone: IK22/5, Biotin, Novus Biologicals™

$252.00 - $329.00

Specifications

Antigen TRANCE/TNFSF11/RANK L
Clone IK22/5
Concentration 0.2mg/mL
Immunogen NSO-derived recombinant mouse TRANCE
Gene ID (Entrez) 8600
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14351501

 
novus biologicals canada
NBP14351501MG
0.1mg Each for $329.00

NBP14351505

 
novus biologicals canada
NBP143515005MG
0.05mg Each for $252.00
Description & Specifications

Specifications

Antigen TRANCE/TNFSF11/RANK L
Clone IK22/5
Concentration 0.2mg/mL
Immunogen NSO-derived recombinant mouse TRANCE
Gene ID (Entrez) 8600
Formulation Phosphate buffer pH 7.2. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry