Please login to your online account to display your discounted pricing

Rat anti-TRAIL/TNFSF10, Clone: N2B2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14327201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14327201

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Mouse
Format Affinity Purified
Isotype IgG2a, κ
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Antigen TRAIL/TNFSF10
Formulation Phosphate buffer pH 7.2
Host Species Rat
Applications Flow Cytometry
Applications Functional Assay
Applications Immunoprecipitation
Immunogen The immunogen for this antibody was Trail.
Purification Method Affinity Purified
Quantity 0.1mg
Primary or Secondary Primary
Gene ID (Entrez) 8743
Clone N2B2
Conjugate Unlabeled