Please login to your online account to display your discounted pricing

Rat anti-TRAIL/TNFSF10, Clone: N2B2, Biotin, Novus Biologicals™

$175.00 - $549.00

Specifications

Antigen TRAIL/TNFSF10
Clone N2B2
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Trail.
Gene ID (Entrez) 8743
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14351601

 
novus biologicals canada
NBP14351601MG
0.1mg Each for $296.00

NBP14351605

 
novus biologicals canada
NBP143516005MG
0.05mg Each for $175.00

NBP1435165

 
novus biologicals canada
NBP14351605MG
0.5mg Each for $549.00
Description & Specifications

Specifications

Antigen TRAIL/TNFSF10
Clone N2B2
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Trail.
Gene ID (Entrez) 8743
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry