Please login to your online account to display your discounted pricing

Rat anti-TLR9, Clone: M9.D6, Novus Biologicals™

$285.00 - $384.00

Specifications

Antigen TLR9
Clone M9.D6
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was Tolllike receptor 9.
Gene ID (Entrez) 54106
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14314101

 
novus biologicals canada
NBP14314101MG
0.1mg Each for $384.00

NBP14314103

 
novus biologicals canada
NBP1431410025MG
0.025mg Each for $285.00
Description & Specifications

Specifications

Antigen TLR9
Clone M9.D6
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was Tolllike receptor 9.
Gene ID (Entrez) 54106
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2a
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Western Blot