Please login to your online account to display your discounted pricing

Rat anti-TLR9, Clone: M9.D6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1431410025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14314103

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Western Blotting
Formulation Phosphate buffer pH 7.2
Primary or Secondary Primary
Gene ID (Entrez) 54106
Storage Requirements Store at 4°C. Do not freeze.
Cross Reactivity Mouse
Conjugate Unlabeled
Antigen TLR9
Concentration 0.2mg/mL
Purification Method Affinity Purified
Immunogen The immunogen for this antibody was Tolllike receptor 9.
Host Species Rat
Quantity 0.025mg
Monoclonal or Polyclonal Monoclonal
Format Affinity Purified
Clone M9.D6
Isotype IgG2a
Regulatory Status RUO