Please login to your online account to display your discounted pricing

Rat anti-TLR9, Clone: M9.D6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14314101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14314101

  • / Each
 

Description & Specifications

Specifications

Antigen TLR9
Applications Flow Cytometry
Applications Western Blot
Clone M9.D6
Concentration 0.2mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Rat
Immunogen The immunogen for this antibody was Tolllike receptor 9.
Isotype IgG2a
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 54106
Target Species Murine