missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-TLR9, Clone: eB72-1665, Novus Biologicals™

$219.00 - $384.00

Specifications

Antigen TLR9
Clone eB72-1665
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Tolllike receptor 9.
Gene Accession No. Q9NR96
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14314001 Novus Biologicals Canada
NBP14314001MG
0.1mg Each for $384.00
NBP14314003 Novus Biologicals Canada
NBP1431400025MG
0.025mg Each for $219.00
Specifications Description & Specifications

Specifications

TLR9
0.5mg/mL
Q9NR96
Phosphate buffer pH 7.2
Primary
Rat
Unlabeled
Affinity Purified
eB72-1665
The immunogen for this antibody was Tolllike receptor 9.
54106
Monoclonal
Store at 4°C. Do not freeze.
IgG2a, κ
Affinity Purified
Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot