Please login to your online account to display your discounted pricing

Rat anti-TLR9, Clone: eB72-1665, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14314001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14314001

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Antigen TLR9
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Rat
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen The immunogen for this antibody was Tolllike receptor 9.
Isotype IgG2a, κ
Purification Method Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Quantity 0.1mg
Gene ID (Entrez) 54106
Clone eB72-1665
Concentration 0.5mg/mL
Conjugate Unlabeled
Gene Accession No. Q9NR96
Cross Reactivity Human