Please login to your online account to display your discounted pricing

Rat anti-TLR6, Clone: hPer6, Novus Biologicals™

$285.00 - $406.00

Specifications

Antigen TLR6
Clone hPer6
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Tolllike receptor 6.
Gene ID (Entrez) 10333
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14314201

 
novus biologicals canada
NBP14314201MG
0.1mg Each for $406.00

NBP14314203

 
novus biologicals canada
NBP1431420025MG
0.025mg Each for $285.00
Description & Specifications

Specifications

Antigen TLR6
Clone hPer6
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Tolllike receptor 6.
Gene ID (Entrez) 10333
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Western Blot