Please login to your online account to display your discounted pricing

Rat anti-TLR6, Clone: hPer6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1431420025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14314203

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Format Affinity Purified
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Immunogen The immunogen for this antibody was Tolllike receptor 6.
Applications Flow Cytometry
Applications Western Blotting
Host Species Rat
Regulatory Status RUO
Formulation Phosphate buffer pH 7.2
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 10333
Primary or Secondary Primary
Isotype IgG2a, κ
Quantity 0.025mg
Antigen TLR6
Clone hPer6
Purification Method Affinity Purified
Concentration 0.5mg/mL