Please login to your online account to display your discounted pricing

Rat anti-TLR6, Clone: hPer6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14314201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14314201

  • / Each
 

Description & Specifications

Specifications

Antigen TLR6
Host Species Rat
Applications Flow Cytometry
Applications Western Blotting
Immunogen The immunogen for this antibody was Tolllike receptor 6.
Isotype IgG2a, κ
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Purification Method Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 10333
Clone hPer6
Concentration 0.5mg/mL
Quantity 0.1mg