Please login to your online account to display your discounted pricing

Rat anti-TIM-3, Clone: 8B.2C12, Biotin, Novus Biologicals™

$197.00 - $549.00

Specifications

Antigen TIM-3
Clone 8B.2C12
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was PE TIM3.
Gene ID (Entrez) 84868
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14353701

 
novus biologicals canada
NBP14353701MG
0.1mg Each for $252.00

NBP14353705

 
novus biologicals canada
NBP143537005MG
0.05mg Each for $197.00

NBP1435375

 
novus biologicals canada
NBP14353705MG
0.5mg Each for $549.00
Description & Specifications

Specifications

Antigen TIM-3
Clone 8B.2C12
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was PE TIM3.
Gene ID (Entrez) 84868
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG1, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry