missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-TIM-1/KIM-1/HAVCR, Clone: RMT1-4, Novus Biologicals™

$285.00 - $329.00

Specifications

Antigen TIM-1/KIM-1/HAVCR
Clone RMT1-4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Purified TIM1.
Gene ID (Entrez) 26762
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP21307901 Novus Biologicals Canada
NBP21307901MG
0.1mg Each for $329.00
NBP21307905 Novus Biologicals Canada
NBP213079005MG
0.05mg Each for $285.00
Specifications Description & Specifications

Specifications

TIM-1/KIM-1/HAVCR
0.5mg/mL
26762
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2b, κ
Affinity Purified
Flow Cytometry
RMT1-4
The immunogen for this antibody was Purified TIM1.
aqueous bufffer may contain carrier protein/stabilizer
Primary
Rat
Unlabeled
Affinity Purified