Please login to your online account to display your discounted pricing

Rat anti-TIM-1/KIM-1/HAVCR, Clone: RMT1-4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP213079005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21307905

  • / Each
 

Description & Specifications

Specifications

Antigen TIM-1/KIM-1/HAVCR
Applications Flow Cytometry
Clone RMT1-4
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation aqueous bufffer may contain carrier protein/stabilizer
Host Species Rat
Immunogen The immunogen for this antibody was Purified TIM1.
Isotype IgG2b, κ
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 26762
Target Species Murine