Please login to your online account to display your discounted pricing

Rat anti-TIM-1/KIM-1/HAVCR, Clone: RMT1-4, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1007764605MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100776465

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Immunogen The immunogen was the full length version of the protein: Tim-1 (RMT1-4)
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Antigen TIM-1/KIM-1/HAVCR
Format Affinity Purified
Host Species Rat
Isotype IgG2b, κ
Purification Method Affinity Purified
Quantity 0.5mg
Gene ID (Entrez) 26762
Clone RMT1-4
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Formulation PBS, pH 7.2. Immunogen affinity purified.