Please login to your online account to display your discounted pricing

Rat anti-Tie-2, Clone: TEK4, PE, Novus Biologicals™

$246.00 - $493.00

Specifications

Antigen Tie-2
Clone TEK4
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was PE Tie2.
Gene ID (Entrez) 7010
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14373301

 
novus biologicals canada
NBP14373301MG
0.1mg Each for $354.00

NBP14373302

 
novus biologicals canada
NBP14373302MG
0.2mg Each for $493.00

NBP14373305

 
novus biologicals canada
NBP143733005MG
0.05mg Each for $246.00
Description & Specifications

Specifications

Antigen Tie-2
Clone TEK4
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was PE Tie2.
Gene ID (Entrez) 7010
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry