Please login to your online account to display your discounted pricing

Rat anti-Thrombomodulin/BDCA-3, Clone: 4D8.2C6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP236496SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP236496SS

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry
Applications Western Blotting
Cross Reactivity Mouse
Quantity 0.025mg
Primary or Secondary Primary
Host Species Rat
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Antigen Thrombomodulin/BDCA-3
Formulation Protein G purified
Conjugate Unlabeled
Gene ID (Entrez) 7056
Immunogen CD141
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Format Purified
Clone 4D8.2C6
Concentration 1.0mg/mL
Purification Method Purified