Please login to your online account to display your discounted pricing

Rat anti-Thrombomodulin/BDCA-3, Clone: 2C7.1E9, Novus Biologicals™

$109.00 - $259.02

Specifications

Antigen Thrombomodulin/BDCA-3
Clone 2C7.1E9
Concentration 1.0mg/mL
Immunogen CD141
Gene ID (Entrez) 7056
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP236497

 
novus biologicals canada
NBP236497
0.1mg Each for $259.02

NBP236497SS

 
novus biologicals canada
NBP236497SS
0.025mg Each for $109.00
Description & Specifications

Specifications

Antigen Thrombomodulin/BDCA-3
Clone 2C7.1E9
Concentration 1.0mg/mL
Immunogen CD141
Gene ID (Entrez) 7056
Formulation Protein G purified
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Rat
Cross Reactivity Mouse
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Western Blotting