Please login to your online account to display your discounted pricing

Rat anti-Thrombomodulin/BDCA-3, Clone: 2C7.1E9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP236497

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP236497

  • / Each
 

Description & Specifications

Specifications

Antigen Thrombomodulin/BDCA-3
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Western Blot
Clone 2C7.1E9
Concentration 1.0mg/mL
Conjugate Unlabeled
Format Purified
Formulation Protein G purified
Host Species Rat
Immunogen CD141
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7056
Target Species Murine