Please login to your online account to display your discounted pricing

Rat anti-TfR (Transferrin R), Clone: R17217, PE, Novus Biologicals™

$246.00 - $493.00

Specifications

Antigen TfR (Transferrin R)
Clone R17217
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was PE CD71.
Gene ID (Entrez) 7037
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14384301

 
novus biologicals canada
NBP14384301MG
0.1mg Each for $354.00

NBP14384302

 
novus biologicals canada
NBP14384302MG
0.2mg Each for $493.00

NBP14384305

 
novus biologicals canada
NBP143843005MG
0.05mg Each for $246.00
Description & Specifications

Specifications

Antigen TfR (Transferrin R)
Clone R17217
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was PE CD71.
Gene ID (Entrez) 7037
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry