Please login to your online account to display your discounted pricing

Rat anti-TfR (Transferrin R), Clone: R17217, FITC, Novus Biologicals™

$246.00 - $708.00

Specifications

Antigen TfR (Transferrin R)
Clone R17217
Immunogen The immunogen for this antibody was CD71.
Gene ID (Entrez) 7037
Formulation PBS [pH 7.2]
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14399101

 
novus biologicals canada
NBP14399101MG
0.1mg Each for $321.00

NBP14399105

 
novus biologicals canada
NBP143991005MG
0.05mg Each for $246.00

NBP1439915

 
novus biologicals canada
NBP14399105MG
0.5mg Each for $708.00
Description & Specifications

Specifications

Antigen TfR (Transferrin R)
Clone R17217
Immunogen The immunogen for this antibody was CD71.
Gene ID (Entrez) 7037
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry