Please login to your online account to display your discounted pricing

Rat anti-TfR (Transferrin R), Clone: R17217, Biotin, Novus Biologicals™

$246.00 - $708.00

Specifications

Antigen TfR (Transferrin R)
Clone R17217
Immunogen The immunogen for this antibody was CD71.
Gene ID (Entrez) 7037
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14361901

 
novus biologicals canada
NBP14361901MG
0.1mg Each for $321.00

NBP14361905

 
novus biologicals canada
NBP143619005MG
0.05mg Each for $246.00

NBP1436195

 
novus biologicals canada
NBP14361905MG
0.5mg Each for $708.00
Description & Specifications

Specifications

Antigen TfR (Transferrin R)
Clone R17217
Immunogen The immunogen for this antibody was CD71.
Gene ID (Entrez) 7037
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry