Please login to your online account to display your discounted pricing

Rat anti-TfR (Transferrin R), Clone: R17217, APC, Novus Biologicals™

$192.00 - $354.00

Specifications

Antigen TfR (Transferrin R)
Clone R17217
Immunogen The immunogen for this antibody was CD71.
Gene ID (Entrez) 7037
Formulation Aqueous buffer (contains stabilizer if necessary)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14303103

 
novus biologicals canada
NBP1430310025MG
0.025mg Each for $192.00

NBP14303101

 
novus biologicals canada
NBP14303101MG
0.1mg Each for $354.00
Description & Specifications

Specifications

Antigen TfR (Transferrin R)
Clone R17217
Immunogen The immunogen for this antibody was CD71.
Gene ID (Entrez) 7037
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate APC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry