Please login to your online account to display your discounted pricing

Rat anti-TfR (Transferrin R), Clone: ER-MP21, FITC, Novus Biologicals™

$536.00 - $536.00

Specifications

Antigen TfR (Transferrin R)
Clone ER-MP21
Concentration 0.1mg/mL
Immunogen ER-MP21 recognizes CD71
Gene ID (Entrez) 7037
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14960401

 
novus biologicals canada
NBP14960401MG
0.1mg Each for $536.00
Description & Specifications

Specifications

Antigen TfR (Transferrin R)
Clone ER-MP21
Concentration 0.1mg/mL
Immunogen ER-MP21 recognizes CD71
Gene ID (Entrez) 7037
Formulation 1mL (100μg/mL) 0.2μm filtered antibody solution in PBS, containing 1% bovine serum. IgG purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a
Conjugate FITC
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)