Please login to your online account to display your discounted pricing

Rat anti-TfR (Transferrin R), Clone: ER-MP21, FITC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP149604005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14960405

  • / Each
 

Description & Specifications

Specifications

Antigen TfR (Transferrin R)
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Formulation 1mL (100μg/mL) 0.2μm filtered antibody solution in PBS, containing 1% bovine serum. IgG purified.
Host Species Rat
Immunogen ER-MP21 recognizes CD71
Isotype IgG2a
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7037
Concentration 0.1mg/mL
Conjugate FITC
Cross Reactivity Mouse
Format Purified
Purification Method Purified
Clone ER-MP21